Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Potassium Dichromate, Reagent Grade
SCI_ED

Catalog No. S001207 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001207 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001207 Supplier Innovating Science™ Supplier No. IS25121
Add to Cart
Add to Cart

Reagent grade potassium dichromate, 500 g, strong oxidizing agent for laboratory, analytical, and educational use with strict safety handling.

  • Potassium Dichromate, Reagent Grade, 500 g - Innovating Science™ is a high-purity inorganic chemical reagent used in laboratory and analytical applications.
  • Specifications:
    • Assay ≥ 99.5%
    • Insoluble Matter ≤ 0.05%
    • Chloride ≤ 0.1%
    • Sulfate ≤ 0.1%
    • Sodium ≤ 0.1%
    • Water ≤ 0.05%
    • Iron ≤ 20 ppm
    • Mercury ≤ 0.1 ppm
  • WARNING : This product can expose you a chemical, Chromium [hexavalent compounds], which is known to the State of California to cause cancer and reproductive harm.

Specifications

Chemical Name or Material Potassium Dichromate
CAS 7778-50-9
Physical Form Crystals
Quantity 500 g
Molecular Weight (g/mol) 294.19
Percent Purity 100%
Grade Reagent
Packaging HDPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.