Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate Solution for UV Visibility, Reagents

Catalog No. 01334216
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-216 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-216 Supplier Reagents Holdings Llc Supplier No. CP202000500C
 Restricted Item
Add to Cart
Add to Cart

Potassium Dichromate Solution for UV Visibility, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Dichromate Solution for UV Visibility
Boiling Point 100°C
Color Orange
CAS 7732-18-5, 7647-01-0, 7778-50-9
Physical Form Liquid
Quantity 500 mL
Molecular Formula K2Cr2O7/H2O
Synonym Potassium bichromate
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0024
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.