Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Dichromate Spectrophotometric Standard, 0.5 AU/cm at 280 nm, Ricca Chemical
SDP

Catalog No. 60875 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6087-5 20 L Cubitainer
6087-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6087-5 Supplier Ricca Chemical Company Supplier No. 60875
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Packaging Cubitainer
CAS Min % 98.0, 0.0045
CAS Max % 100.0, 0.0047
Physical Form Liquid
Color Orange
Boiling Point 100°C
Quantity 20 L
Chemical Name or Material Potassium Dichromate Spectrophotometric Standard, 0.5AU/cm at 28 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.