Learn More
Crescent Chemical Co Inc POTASSIUM DIHYDROGEN PHOSPHATE
SDP
Catalog No. 50247764
Encompass_Preferred
Potassium dihydrogen phosphate anhydrous analytical grade, Ph. Eur. Biochemical and enzyme standard. Buffering substance according to Sörensen. 9.072 g in 1 liter water = 1/15 M. Tested for use in tissue culture. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Numb
Catalog No. 50-247-764 Supplier Crescent Chemical Co Inc Supplier No. SE26870.01
Potassium dihydrogen phosphate anhydrous analytical grade, Ph. Eur. Biochemical and enzyme standard. Buffering substance according to Sörensen. 9.072 g in 1 liter water = 1/15 M. Tested for use in tissue culture. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Numb