Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Disulfate

Catalog No. 307750500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-775-0500G 500 g
30-775-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 30-775-0500G Supplier Sigma Aldrich Supplier No. 307750500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium disulfate (K2S2O7) can be used: As a salt catalyst for the conversion of hemicellulose into reducing sugars such as xylose and galactose by mechanocatalysis . In the preparation of diacylbenzenes via Pd-catalyzed decarboxylative acylation of aromatic ketones with oxocarboxylic acids. As an oxidant for the preparation of monosubstituted triazoles from aryl azides and propargyl alcohol using Ag2O as a catalyst.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 7790-62-7
Quantity 500 g
Molecular Formula K2S2O7
Linear Formula K2S2O7
MDL Number MFCD00011385
Synonym Potassium pyrosulfate
Molecular Weight (g/mol) 254.32 g/mol
Density 2.28 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.