Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Disulfate, EMSURE™, MilliporeSigma™,

Catalog No. M1051079050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Plastic Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1051079050 50 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1051079050 Supplier MilliporeSigma Supplier No. 1051079050
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
Synonym potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat
PubChem CID 62681
IUPAC Name dipotassium;sulfonato sulfate
SMILES [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 325°C
Color White
pH 0.8 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 50 kg
Formula Weight 254.33
Percent Purity 97.5 to 100.0%
Grade ACS
Packaging Plastic Drum
Chemical Name or Material Potassium Pyrosulfate
EINECSNumber 232-216-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.