Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Disulfate, Puriss. p.a. ≥97.5% (T), Solstice

Catalog No. 60047169 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-169 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-169 Supplier Solstice Supplier No. 602351KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥97.5% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
MDL Number MFCD00011385
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
Synonym potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat
PubChem CID 62681
IUPAC Name dipotassium;sulfonato sulfate
SMILES [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 325°C
Color White
pH 1.0 to 2.0 (25.5g/L at 25°C)
Physical Form Solid
Quantity 1 kg
UN Number UN3260
Formula Weight 254.32g/mol
Percent Purity ≥97.5% (T)
Packaging Poly bottle
Density 2.28g/mL at 25°C
Chemical Name or Material Potassium disulfate
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
DOTInformation 8/PGII
RTECSNumber 232-216-8
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.