Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium disulfitopalladate(II), Premion™, 99.95% (metals basis), Pd 30.4% min, Thermo Scientific™

Catalog No. AA4101106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4101106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4101106 Supplier Thermo Scientific Chemicals Supplier No. 4101106

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68310-13-4
Molecular Formula KO3PdS
Molecular Weight (g/mol) 225.58
MDL Number MFCD00792331
InChI Key FULLCBDUCZPCNB-UHFFFAOYSA-L
Synonym Potassium palladium sulfite
IUPAC Name potassium palladium sulfite
SMILES [K+].[Pd].[O-]S([O-])=O

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 344.75
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 30.4% min.
Assay (metals basis)
Chemical Name or Material Potassium disulfitopalladate(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 269-702-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.