Learn More
World Precision Instrument Potassium Electrode Tips, Replacement
SDP
Catalog No. 502521042
Encompass
Replacement tips
Set of 3 potassium replacement tips for ion selective electrodes. Change the tip on your holder in less than a minute. Can be stored 3-4 months under refrigeration.
Catalog No. 50-252-1042 Supplier World Precision Instrument Supplier No. TIPK
Replacement tips
Set of 3 potassium replacement tips for ion selective electrodes. Change the tip on your holder in less than a minute. Can be stored 3-4 months under refrigeration.