Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Ferricyanide Solution, 1%, Fisher Scientific™
SDP

Catalog No. SP9401250 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP9401250 250 mL Natural Poly Bottle
SP9401-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP9401250
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13746-66-2 , 7732-18-5
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Percent Purity 1%
Grade Laboratory
Packaging Natural Poly Bottle
Methods For AOAC Method 953.08
Quantity 250 mL
Chemical Name or Material Potassium Ferricyanide Solution, 1%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.