Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Ferrocyanide, Trihydrate, Crystal, Reagent, ACS, 98.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18605548 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-548 25 g Amber Glass Bottle
18-605-549 125 g Amber Glass Bottle
18-605-550 500 g Amber Glass Bottle
18-605-552 2.5 kg Amber Glass Bottle
18-605-551 6 x 500 g Amber Glass Bottle
18-605-553 4 x 2.5 kg Amber Glass Bottle
18-605-554 12 kg Poly Pail
18-605-555 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-605-548 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P129525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1295, 14459-95-1, K4Fe(CN)6·3H2O

Spectrum™ Chemical Potassium Ferrocyanide, Trihydrate, Crystal, Reagent, ACS is an inorganic compound that is a salt with a light yellow tint. Mainly used as an anti-caking agent in both road and table salt. It has several other niche uses and within the laboratory to determine potassium permanganate concentration levels. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98.5 to 102%
Grade ACS
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.