Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, 40% (w/v), Ricca Chemical
SDP

Catalog No. 61045 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6104-5 20 L Cubitainer
6100-16 500 mL Natural Poly Bottle
6100-32 1 L Natural Poly Bottle
61001 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6104-5 Supplier Ricca Chemical Company Supplier No. 61045
Add to Cart
Add to Cart

For APHA/ASTM/EPA Testing

Chemical Identifiers

Molecular Formula FK
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Color Colorless to Pink
CAS 7789-23-3, 7732-18-5
CAS Min % 66.0, 31.67
CAS Max % 69.0, 32.96
Molecular Weight (g/mol) 58.10
Percent Purity 40%
Grade Laboratory
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
pH ∼7
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 40% (w/w)
Chemical Name or Material Potassium Fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.