Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride Dihydrate, Reagent Grade, Reagents

Catalog No. 01337844
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-844 500 g
01-337-845 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-844 Supplier Reagents Holdings Llc Supplier No. C1284000500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Fluoride Dihydrate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
IUPAC Name potassium dihydrate fluoride
SMILES O.O.[F-].[K+]

Specifications

Boiling Point 156°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.45
Chemical Name or Material Potassium Fluoride Dihydrate, Reagent Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.