Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, For Dissolved Oxygen, Certified, 40% (w/v), LabChem™

Catalog No. p-5389177
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC191001 500 mL
LC191002 1 L
LC191004 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC191001 Supplier LabChem, Inc. Supplier No. LC191001
Only null left
Add to Cart
Add to Cart

Specifications

Color Clear, colorless
Format Liquid
Molecular Formula FK
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Packaging Poly Bottle
SMILES [F-].[K+]
IUPAC Name potassium fluoride
Molecular Weight (g/mol) 58.10
Quantity 500 mL
Storage Requirements Room Temperature
Type Potassium Fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.