Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride Solution, 3 Molar, Reagents

Catalog No. 01336184
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-184 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-184 Supplier Reagents Holdings Llc Supplier No. C84007NE1A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Fluoride Solution, 3 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Fluoride Solution, 3 Molar
Boiling Point 103°C
CAS 7732-18-5, 13455-21-5, 13455-21-5
Color Colorless
Physical Form Liquid
Quantity 1 L
Molecular Formula KF/H2O
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1401
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.