Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Fluoride Solution, 3 Molar, Reagents
Click to view available options
Quantity:
1 L
4 L
Specifications
Specifications
| Chemical Name or Material | Potassium Fluoride Solution, 3 Molar |
| Boiling Point | 103°C |
| CAS | 7732-18-5, 13455-21-5, 13455-21-5 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 1 L |
| Molecular Formula | KF/H2O |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.1401 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction