Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride Solution, NMR Reference Standard, 0.2 M in Water, D2O 10% (99.9 atom % D), MilliporeSigma™ Supelco™

Catalog No. 111002601 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1.9 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2601 10 x 1.9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2601 Supplier MilliporeSigma Supelco Supplier No. 61287110X8IN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NMR Tube Size: 8 mm 8 in.

TRUSTED_SUSTAINABILITY

Specifications

Packaging NMR Tube Size: 8 mm 8 in.
Concentration 0.2 M aq. soln.
MDL Number MFCD00011398
Quantity 10 x 1.9 mL
Grade NMR Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.