Learn More
Strem, An Ascensus Company CAS# 12081-88-8. 5g. Potassium graphite KC8. MFCD01324308
SDP
Supplier:  Strem, An Ascensus Company 1919355G
Encompass
CAS# 12081-88-8. 5g. Potassium graphite KC8. MFCD01324308. Molecular Weight: 135.18. Molecular Formula: C8K. Color/form: bronze solid Strem# 19-1935. http://www.strem.com/catalog/v/19-1935/
Catalog No. NC1015155