Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium heptafluorotantalate(V), 99.99% (metals basis)

Catalog No. p-7058996
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4373309 10 g
AA4373318 50 g
AA4373330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4373309 Supplier Thermo Scientific Chemicals Supplier No. 04373309
Only null left
Add to Cart

CAS: 16924-00-8 | F7K2Ta | 392.133 g/mol

It is an important raw material used as a primary and secondary pharmaceutical intermediate and also used in the production and preparation of tantalum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16924-00-8
Molecular Formula F7K2Ta
Molecular Weight (g/mol) 392.133
MDL Number MFCD00042369
InChI Key DPXAUKPQZOAHGF-UHFFFAOYSA-G
Synonym potassium heptafluorotantalate, potassium tantalum fluoride, dipotassium heptafluorotantalate, tantalum potassium fluoride, potassium fluotantalate, potassium heptafluorotantalate v, potassium fluorotantalate, dipotassium heptafluorotantalum 2-, potassium fluorotanatalate k2taf7, potassium tantalum fluoride k2taf5
PubChem CID 28146
IUPAC Name dipotassium;heptafluorotantalum(2-)
SMILES F[Ta-2](F)(F)(F)(F)(F)F.[K+].[K+]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information Decomposes in water. Slightly soluble in hydrogen fluoride.
Formula Weight 392.14
Percent Purity 99.99%
Density 4.560 g/mL
Assay (metals basis)
Chemical Name or Material Potassium heptafluorotantalate (V)
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 240-986-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.