Learn More
Strem, An Ascensus Company CAS# 16920-56-2. 5g. Potassium hexachloroiridate(IV), 99%. MFCD00011369
SDP
Catalog No. 5090113160
Encompass
CAS# 16920-56-2. 5g. Potassium hexachloroiridate(IV), 99%. MFCD00011369. Molecular Weight: 483.12. Molecular Formula: K2IrCl6. Color/form: black pwdr. Strem# 77-6500. http://www.strem.com/catalog/v/77-6500/
Catalog No. 50-901-13160 Supplier Strem, An Ascensus Company Supplier No. 7765005G
CAS# 16920-56-2. 5g. Potassium hexachloroiridate(IV), 99%. MFCD00011369. Molecular Weight: 483.12. Molecular Formula: K2IrCl6. Color/form: black pwdr. Strem# 77-6500. http://www.strem.com/catalog/v/77-6500/