Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Hexachloroplatinate(Iv)

Catalog No. 2060671G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-606-71G 1 g
20-606-7100G 100 g
20-606-725G 25 g
20-606-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 20-606-71G Supplier Sigma Aldrich Supplier No. 2060671G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium Hexachloroplatinate(Iv)

Specifications

CAS 16921-30-5
Melting Point 250°C
Molecular Formula Cl6K2Pt
Linear Formula K2PtCl6
Quantity 1 g
MDL Number MFCD00011389
Synonym Dipotassium hexachloroplatinate; Dipotassium platinum hexachloride; Platinic potassium chloride; Platinum potassium chloride; Potassium chloroplatinate; Potassium platinum(IV) chloride; Potassium platinum(IV) hexachloride
Molecular Weight (g/mol) 485.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.