Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min

Catalog No. AA4039903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4039903 1 g
AA4039906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4039903 Supplier Thermo Scientific Chemicals Supplier No. 04039903
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23013-82-3 | K2RuCl6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23013-82-3
Molecular Formula K2RuCl6
MDL Number MFCD00050159

Specifications

Color Red to Brown
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 99.95% (metals basis)
Formula Weight 391.99
Appearance Red-browm crystalline powder
Chemical Name or Material Potassium hexachlororuthenate(IV)
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-381-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.