Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hexacyanoferrate(Ii) Trihydrate, purum, Solstice

Catalog No. 6001953 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-53 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-019-53 Supplier Solstice Supplier No. 1263925KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Physical Form Crystalline
Quantity 25 kg
Linear Formula K4Fe(CN)6 · 3H2O
UN Number NONH for all modes of transport
Formula Weight 422.39g/mol
Percent Purity ≥99%
Grade purum
Packaging Poly Drum
Chemical Name or Material Potassium Hexacyanoferrate(Ii) Trihydrate
Precautionary Statement H412
EINECSNumber 237-722-2
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.