Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hexacyanoferrate(III), 99%, ReagentPlus™, Solstice

Catalog No. 60048146 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-146 500 g Poly bottle
60-048-145 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-146 Supplier Solstice Supplier No. P8131500G

Reagent Grade, ∼99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium;iron(3+);hexacyanide
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Red
pH 8.0 (at 20°C)
Physical Form Crystalline
Quantity 500 g
Linear Formula K3Fe(CN)6
Formula Weight 329.24g/mol
Percent Purity 99%
Odor Odorless
Packaging Poly bottle
Density 1.85g/cu.cm at 20°C
Chemical Name or Material Potassium hexacyanoferrate(III)
Show More Show Less
RTECSNumber 237-323-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.