Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hexacyanoferrate(III), For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1049731000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1049731000 1 kg Plastic Bottle
M1049730100 100 g Plastic Bottle
M1049730250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1049731000 Supplier MilliporeSigma Supplier No. 1049731000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Red
pH 6 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 1 kg
Solubility Information 464g/L
Formula Weight 329.25
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 1.85 g/3 (20 °C)
EINECSNumber 237-323-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.