Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Potassium Hydroxide 0.5N in Benzyl Alcohol (4L)
SDP

Catalog No. NC0259250
Encompass_Preferred

Potassium Hydroxide 0.5N in Benzyl Alcohol (4L)

Catalog No. NC0259250 Supplier Aqua Solutions Supplier No. HUN007
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Hydroxide 0.5N in Benzyl Alcohol (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.