Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydride, 30 wt.% Dispersion in Mineral Oil, Thermo Scientific™

Catalog No. AC329481000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC329481000 100 g
AC329480250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329481000 Supplier Thermo Scientific Chemicals Supplier No. 329481000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7693-26-7
Molecular Formula HK
Molecular Weight (g/mol) 40.106
InChI Key OCFVSFVLVRNXFJ-UHFFFAOYSA-N
Synonym potassium hydride, potassium hydride kh, kaliumhydrid, acmc-20ajuo, kh, potassium hydride w/w in mineral oil
PubChem CID 82127
IUPAC Name potassium;hydride
SMILES [H-].[K+]

Specifications

Color Tan
Concentration 29 to 31%
Density 1.45
Formula Weight 40.11
Quantity 100 g
Physical Form Dispersion of owder In Mineral Oil
Chemical Name or Material Potassium hydride
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-704-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.