Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Hydride

Catalog No. 70886025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-886-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 70-886-025G Supplier Sigma Aldrich Supplier No. 70886025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium hydride is a strong base and hydride donor to Lewis acids. It is generally employed in organic reactions like deprotonation, rearrangement, and elimination reactions. It is also used as a reducing agent to reduce hindered boranes and borates to substituted borohydrides.Potassium hydride in paraffin can be used as a base in:Wittig reaction of various aldehydes with high Z selectivity.Williamson ether synthesis to prepare substituted ethers from alcohols.Polymerization of ethylene oxide to yield polyethylene glycols.

Specifications

CAS 7693-26-7
Quantity 25 g
Molecular Formula HK
Linear Formula KH
MDL Number MFCD00011357
Molecular Weight (g/mol) 40.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.