Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydrogen phosphate, dried, 98+%

Catalog No. AAA113210I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA113210I 5000 g
AAA1132130 250 g
AAA113210B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA113210I Supplier Thermo Scientific Chemicals Supplier No. A113210I
Only null left
Add to Cart

CAS: 7758-11-4 | HK2O4P | 174.17 g/mol

Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic chemical and incompatible with strong oxidizing agents and acids. Keep the material away from heat, sparks and flame and sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Quantity 5000 g
Merck Index 14,7658
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 174.18
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material Potassium hydrogen phosphate, Dried
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 231-834-5
RTECSNumber TC5580000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.