Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium hydrogen phthalate, 877-24-7, MFCD00013070, 60 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NIST84L
Encompass
Linear Formula: HOOCC6H4COOK, Molecular Weight: 204.22, NIST SRM 84l, Synonym: KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate, Potassium phthalate monobasic.
Catalog No. NC1533962