Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Sulfate ACS AR Powder, Macron Fine Chemicals™

Catalog No. MK7112500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-7112-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7112-500 Supplier Avantor Supplier No. 711204
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Specifications

CAS 7646-93-7
Physical Form Powder
Quantity 500 g
Molecular Formula HKO4S
MDL Number MFCD00011404
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
SMILES [K+].OS([O-])(=O)=O
IUPAC Name potassium hydrogen sulfate
Molecular Weight (g/mol) 136.16
PubChem CID 516920
Formula Weight 136.17
Percent Purity >95%
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Potassium Hydrogen Sulfate, Fused
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.