Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Sulfate, Fused, ACS Reagent, Reagents

Catalog No. 01337793
Change view
Click to view available options
Quantity:
500 g
3 kg
Catalog No. Quantity
01-337-793 500 g
01-337-794 3 kg
2 options

Catalog No. 01-337-793

Supplier: Reagents Holdings Llc C1290200500C2

Potassium Hydrogen Sulfate, Fused, ACS Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
Synonym Potassium Hydrogen Sulfate
IUPAC Name dipotassium (sulfonatooxy)sulfonate
SMILES [K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1
Chemical Name or Material Potassium Hydrogen Sulfate, Fused, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.