Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Sulfate, Fused, ACS Reagent, Reagents

Catalog No. 01337794
Change view
Click to view available options
Quantity:
3 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-794 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-794 Supplier Reagents Holdings Llc Supplier No. C12902003A5

missing translation for 'validMainframeMsg'

Potassium Hydrogen Sulfate, Fused, ACS Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
Synonym Potassium Hydrogen Sulfate
IUPAC Name dipotassium (sulfonatooxy)sulfonate
SMILES [K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1
Chemical Name or Material Potassium Hydrogen Sulfate, Fused, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.