Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Sulfate, MP Biomedicals

Catalog No. ICN15634090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15634090 500 g
ICN15634083 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15634090 Supplier MP Biomedicals, Inc Supplier No. 0215634090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7646-93-7 | Formula: HKO4S | Weight: 136.16 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Quantity 500 g
Formula Weight 136.2
Chemical Name or Material Potassium Hydrogen Sulfate
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.