Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Hydrogenfluoride Solution

Catalog No. 663883100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-388-3100ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 66-388-3100ML Supplier Sigma Aldrich Supplier No. 663883100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used to prepare potassium organotrifluoroborates from their corresponding boronic acids.

Specifications

CAS 99770-93-1
Melting Point 241°C to 245°C
Molecular Formula C18H28B2O4
Linear Formula C18H28B2O4
MDL Number MFCD08276852
Quantity 5 g
Synonym Potassium acid bifluoride solution; Potassium acid fluoride solution; Potassium bifluoride; Potassium fluoride hydrofluoride solution; Potassium hydrogen difluoride; Potassium monohydrogen difluoride solution
Molecular Weight (g/mol) 330.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.