Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions POTASSIUM HYDROXIDE 0.01N AQ
SDP

Catalog No. NC0248122
Encompass_Preferred

POTASSIUM HYDROXIDE 0.01N AQ ..

Catalog No. NC0248122 Supplier Aqua Solutions Supplier No. 71524L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

POTASSIUM HYDROXIDE 0.01N AQ ..

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.