Learn More
Ethos Biosciences Potassium Hydroxide 10PCT DMSO
Catalog No. 5000819183
Encompass_Preferred
Potassium Hydroxide 10% with DMSO is a ready-to-use alkaline solution suitable for pathology cytology microbiology and general laboratory applications Commonly used in microbiology for KOH preparations fungal detection and specimen clearing of skin hair and nail samples it is ideal for routine diagnostic procedures The clear colorless solution provides consistent performance for reliable microscopic evaluation and analytical tasks It is also appropriate for tissue digestion pH adjustment cleaning and general chemical processing Supplied in chemically compatible containers Potassium Hydroxide 10% with DMSO is suitable for clinical research pharmaceutical and industrial laboratories
Catalog No. 50-008-19183 Supplier Ethos Biosciences Supplier No. 3330D02
Potassium Hydroxide 10% with DMSO is a ready-to-use alkaline solution suitable for pathology cytology microbiology and general laboratory applications Commonly used in microbiology for KOH preparations fungal detection and specimen clearing of skin hair and nail samples it is ideal for routine diagnostic procedures The clear colorless solution provides consistent performance for reliable microscopic evaluation and analytical tasks It is also appropriate for tissue digestion pH adjustment cleaning and general chemical processing Supplied in chemically compatible containers Potassium Hydroxide 10% with DMSO is suitable for clinical research pharmaceutical and industrial laboratories