Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 40% (w/w), Ricca Chemical
SDP

Catalog No. 61805 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61805 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61805 Supplier Ricca Chemical Company Supplier No. 61805
Add to Cart
Add to Cart

Specifications

Color Colorless
CAS 1310-58-3, 7732-18-5
CAS Min % 54.0, 43.79
CAS Max % 56.0, 45.57
Molecular Formula HKO
MDL Number MFCD00003553
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Percent Purity 40%
Grade Laboratory
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
pH >13
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 40% (w/w)
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.