Learn More
Supply Solutions Avantor Performance Materials - Potassium Hydroxide, Pellets. Size: 12Kg; Grade: Ar (Acs); Assay: 85%
Catalog No. 50878134
Encompass_Preferred
Avantor Performance Materials - Potassium Hydroxide, Pellets. Size: 12kg; Grade: AR (ACS); Assay: 85%.; Size: 12kg; Grade: AR (ACS); Assay: 0.85; CAS: 1310-58-3; Hazardous: Y; Container Material: STAKMOR; Formula: KOH; Formula Weight: 56.11
Catalog No. 50-878-134 Supplier Supply Solutions Supplier No. MACRON 698420
Avantor Performance Materials - Potassium Hydroxide, Pellets. Size: 12kg; Grade: AR (ACS); Assay: 85%.; Size: 12kg; Grade: AR (ACS); Assay: 0.85; CAS: 1310-58-3; Hazardous: Y; Container Material: STAKMOR; Formula: KOH; Formula Weight: 56.11