Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium hydroxide ACS re25KG
Catalog No. 502897447
Encompass
Potassium hydroxide is an inorganic salt mainly used as a precursor to synthesize other potassium compounds such as potassium acetate potassium bicarbonate and potassium carbonate. On a commercial scale it is synthesized by the electrolysis of potassium chloride.
Catalog No. 50-289-7447 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2214732.5KG
Potassium hydroxide is an inorganic salt mainly used as a precursor to synthesize other potassium compounds such as potassium acetate potassium bicarbonate and potassium carbonate. On a commercial scale it is synthesized by the electrolysis of potassium chloride.