Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Concentrate 0.1 M KOH in Water (0.1 N), Eluent concentrate for IC, MilliporeSigma™ Supelco™

Catalog No. 111020299 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0299 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0299 Supplier MilliporeSigma Supelco Supplier No. 616991L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Prepared with potassium hydroxide and high purity water (18.2 MO mega, 0.2 μm filtered)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3, 7732-18-5
Linear Formula KOH
MDL Number MFCD00003553
Synonym Potassium hydroxide solution
Quantity 1 L
Molecular Weight (g/mol) 56.11
Formula Weight 56.11
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.