Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium hydroxide free-f1KG
Catalog No. 502899227
Encompass
Potassium hydroxide is an inorganic salt mainly used as a precursor to synthesize other potassium compounds such as potassium acetate potassium bicarbonate and potassium carbonate. On a commercial scale it is synthesized by the electrolysis of potassium chloride.
Catalog No. 50-289-9227 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9110621KG
Add to Cart
Add to Cart
Potassium hydroxide is an inorganic salt mainly used as a precursor to synthesize other potassium compounds such as potassium acetate potassium bicarbonate and potassium carbonate. On a commercial scale it is synthesized by the electrolysis of potassium chloride.