Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomas Scientific Potassium Hydroxide in Meth 0 50N 20L UN2924
SDP

Catalog No. 5000550666
Encompass_Preferred

Potassium Hydroxide in Meth 0 50N 20L UN2924

Catalog No. 50-005-50666 Supplier Thomas Scientific Supplier No. C748B91
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Hydroxide in Meth 0 50N 20L UN2924

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.