Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide NF GenAR Pellets, Macron Fine Chemicals™

Catalog No. MK7815500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 12 x 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-7815-500 POLYSTORMOR; 12 x 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7815-500 Supplier Avantor Supplier No. 781504

missing translation for 'validMainframeMsg'

Specifications

CAS 1310-58-3
Molecular Formula HKO
MDL Number MFCD00003553
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Quantity 12 x 500 g
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11
Grade NF
Packaging POLYSTORMOR
Physical Form Pellet
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.