Learn More
Strem, An Ascensus Company CAS# 1310-58-3. 500g. Potassium hydroxide, pellets, 85+% (ACS). MFCD00003553
SDP
Catalog No. 5090110633
Encompass
CAS# 1310-58-3. 500g. Potassium hydroxide, pellets, 85+% (ACS). MFCD00003553. Molecular Weight: 56.11. Molecular Formula: KOH. Color/form: white pellets Strem# 19-3150. http://www.strem.com/catalog/v/19-3150/
Catalog No. 50-901-10633 Supplier Strem, An Ascensus Company Supplier No. 193150500G
CAS# 1310-58-3. 500g. Potassium hydroxide, pellets, 85+% (ACS). MFCD00003553. Molecular Weight: 56.11. Molecular Formula: KOH. Color/form: white pellets Strem# 19-3150. http://www.strem.com/catalog/v/19-3150/