Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide/Potassium Cyanide Solution, AOAC 983.19, Reagents

Catalog No. 01334118
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-118 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-118 Supplier Reagents Holdings Llc Supplier No. CP2600001A
 Restricted Item
Add to Cart
Add to Cart

Potassium Hydroxide/Potassium Cyanide Solution, AOAC 983.19, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Hydroxide/Potassium Cyanide Solution, AOAC 983.19
Color Colorless
CAS 7732-18-5, 1310-58-3, 151-50-8
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1653
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.