Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium hydroxide soluti
Catalog No. 502898606
Encompass
Potassium hydroxide (KOH) SOLUTIONon is a strong alkali widely used in chemical synthesis. Van Slyke apparatus and Hildebrand adsorption technique have been utilized to determine the solubility of oxygen in different concentration of KOH SOLUTIONon.
Catalog No. 50-289-8606 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 41766118L
Potassium hydroxide (KOH) SOLUTIONon is a strong alkali widely used in chemical synthesis. Van Slyke apparatus and Hildebrand adsorption technique have been utilized to determine the solubility of oxygen in different concentration of KOH SOLUTIONon.