Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioworld Potassium Hydroxide Solution 50% (w/v), 4 L

Catalog No. 50255554
Encompass_Preferred

Potassium Hydroxide Solution, 50% (w/v) in waterHaz Mat Fee Applies

Catalog No. 50-255-554 Supplier Bioworld Supplier No. 402201223
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Hydroxide Solution, 50% (w/v) in waterHaz Mat Fee Applies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.