Learn More
Bioworld Potassium Hydroxide Solution 1.0N, 500 mL
Catalog No. 50255558
Encompass_Preferred
Potassium Hydroxide, 1.0N Solution in waterUseful for HPLCHaz Mat Fee AppliesAppearance: LiquidBoiling Point : 1320 °CDensity : 1.450 g/mL at 20 °C
Catalog No. 50-255-558 Supplier Bioworld Supplier No. 402201241
Potassium Hydroxide, 1.0N Solution in waterUseful for HPLCHaz Mat Fee AppliesAppearance: LiquidBoiling Point : 1320 °CDensity : 1.450 g/mL at 20 °C