Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide Solution, 0.05N (In Methanol/Certified), Fisher Chemical

Catalog No. DW806181 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DW806181 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DW806181

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
Boiling Point 66.1°C
Melting Point -100°C
Concentration 0.05N
UN Number 2924
Density 0.8
Solubility Information Soluble in water
Quantity 1 L
Odor Alcohol-like
Physical Form Liquid
DOTInformation Hazard Class 3; Packing Group II
Recommended Storage Flammables area. Keep container tightly closed in a dry and well-ventilated place. Keepaway from open flames, hot surfaces and sources of ignition.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.